accessibility menu, dialog, popup

UserName

CH3H2NH3CF

CAS RN 392-70-1

IUPAC Name: 4-fluoro-2,6-dimethylaniline
Synonyms: 4-fluoro-2,6-dimethylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: MPNDLCBMLBACDD-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)