accessibility menu, dialog, popup

UserName

CAS RN 38894-11-0

IUPAC Name: (2Z)-2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole
Synonyms: (2Z)-2-hydrazinylidene-3-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 179.24
Molecular Formula: C8H9N3S
InChi Key: PHOLIFLKGONSGY-NTMALXAHSA-N
SMILES: CN1\C(SC2=CC=CC=C12)=N\N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)