accessibility menu, dialog, popup

UserName

OCNNCO

CAS RN 38661-72-2

IUPAC Name: 1,3-bis(isocyanatomethyl)cyclohexane
Synonyms: 1,3-bis(isocyanatomethyl)cyclohexane
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: XSCLFFBWRKTMTE-UHFFFAOYNA-N
SMILES: O=C=NCC1CCCC(CN=C=O)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results