accessibility menu, dialog, popup

UserName

H3C

CAS RN 38641-16-6

IUPAC Name: 1-dodecylnaphthalene
Synonyms: 1-dodecylnaphthalene
Molecular Weight (g/mol): 296.5
Molecular Formula: C22H32
InChi Key: UVMGANPLBFFQIO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results