accessibility menu, dialog, popup

UserName

OSOOFFF

CAS RN 3857-83-8

IUPAC Name: naphthalen-2-yl trifluoromethanesulfonate
Synonyms: naphthalen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 276.23
Molecular Formula: C11H7F3O3S
InChi Key: MDWRQYBWVTXIIJ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)