accessibility menu, dialog, popup

UserName

H3CNH2NH2N

CAS RN 38483-26-0

IUPAC Name: 5-ethyl-6-phenyl-5H,6H-benzo[h]isoquinoline-4,8-diamine
Synonyms: 5-ethyl-6-phenyl-5H,6H-benzo[h]isoquinoline-4,8-diamine
Molecular Weight (g/mol): 315.42
Molecular Formula: C21H21N3
InChi Key: ZDWPSSBKIMIPMY-UHFFFAOYNA-N
SMILES: CCC1C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CN=CC(N)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results