accessibility menu, dialog, popup

UserName

CAS RN 38465-86-0

IUPAC Name: iridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide bis(methyldiphenylphosphane)
Synonyms: iridium(1+) 1,5-cyclooctadiene, (Z,Z)- bis(methyldiphenylphosphane) hexafluorophosphate
Molecular Weight (g/mol): 845.81
Molecular Formula: C34H38F6IrP3
InChi Key: LXKHQEQLBSJJCO-JXNOXZOESA-N
SMILES: [Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)