accessibility menu, dialog, popup

UserName

CAS RN 3843-74-1

IUPAC Name: methyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms: methyl caffeate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: OCNYGKNIVPVPPX-HWKANZROSA-N
SMILES: COC(=O)\C=C\C1=CC=C(O)C(O)=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)