missing translation for 'onlineSavingsMsg'
Learn More

Methyl caffeate

Catalog No. AAJ63786MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63786MD 250 mg
AAJ6378603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63786MD Supplier Thermo Scientific Chemicals Supplier No. J63786MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of protein tyrosine kinase | CAS: 3843-74-1 | C10H10O4 | 194.186 g/mol

Inhibitor of protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3843-74-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00210468
InChI Key OCNYGKNIVPVPPX-HWKANZROSA-N
Synonym methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001
PubChem CID 689075
ChEBI CHEBI:6856
IUPAC Name methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Melting Point 158°C to 161°C
Color Yellow
Quantity 250 mg
Merck Index 14,1635
Formula Weight 194.19
Physical Form Powder
Chemical Name or Material Methyl caffeate
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only