missing translation for 'onlineSavingsMsg'
Learn More

Methyl Caffeate 98.0+%, TCI America™

Catalog No. M2519200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2519200MG 200 mg
M25191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2519200MG Supplier TCI America Supplier No. M2519200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3843-74-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00210468
InChI Key OCNYGKNIVPVPPX-HWKANZROSA-N
Synonym methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001
PubChem CID 689075
ChEBI CHEBI:6856
IUPAC Name methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Melting Point 160°C
Quantity 200 mg
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Caffeate