accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 38409-59-5

IUPAC Name: 1-bromo-3-hexylbenzene
Synonyms: 1-bromo-3-hexylbenzene
Molecular Weight (g/mol): 241.17
Molecular Formula: C12H17Br
InChi Key: HZUVRCRPECDFAT-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC(Br)=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)