missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-n-hexylbenzene, 97+%

Catalog No. AAH2606314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2606314 25 g
AAH2606306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2606314 Supplier Thermo Scientific Chemicals Supplier No. H2606314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38409-59-5 | C12H17Br | 241.172 g/mol

1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

Solubility
Insoluble in water. Fully miscible with ethanol, acetone, toluene, methylene chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 38409-59-5
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD09909567
InChI Key HZUVRCRPECDFAT-UHFFFAOYSA-N
Synonym 1-bromo-3-n-hexylbenzene, 1-bromo-3-hexyl-benzene, 3-hexylbromobenzene, 3-n-hexyl bromobenzene, #, benzene, 1-bromo-3-hexyl
PubChem CID 563022
IUPAC Name 1-bromo-3-hexylbenzene
SMILES CCCCCCC1=CC(=CC=C1)Br

Specifications

Density 1.179
Boiling Point 90°C to 98°C (5 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.522
Quantity 25 g
Solubility Information Insoluble in water. Fully miscible with ethanol,acetone,toluene,methylene chloride.
Formula Weight 241.17
Percent Purity ≥97%
Chemical Name or Material 1-Bromo-3-n-hexylbenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only