missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-hexylbenzene 98.0+%, TCI America™

Catalog No. B41805G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41805G Supplier TCI America Supplier No. B41805G

Chemical Identifiers

CAS 38409-59-5
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD09909567
InChI Key HZUVRCRPECDFAT-UHFFFAOYSA-N
Synonym 1-bromo-3-n-hexylbenzene, 1-bromo-3-hexyl-benzene, 3-hexylbromobenzene, 3-n-hexyl bromobenzene, #, benzene, 1-bromo-3-hexyl
PubChem CID 563022
IUPAC Name 1-bromo-3-hexylbenzene
SMILES CCCCCCC1=CC(=CC=C1)Br

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
Formula Weight 241.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-hexylbenzene