accessibility menu, dialog, popup

UserName

BrBr

CAS RN 38399-13-2

IUPAC Name: 1-bromo-2-[2-(2-bromophenyl)ethynyl]benzene
Synonyms: 1-bromo-2-[2-(2-bromophenyl)ethynyl]benzene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: RVGSIXSCGKMAOK-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C#CC1=CC=CC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results