accessibility menu, dialog, popup

UserName

CAS RN 3839-35-8

H3COSOOCH3

CAS RN 3839-35-8

IUPAC Name: hexyl 4-methylbenzene-1-sulfonate
Synonyms: hexyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 256.36
Molecular Formula: C13H20O3S
InChi Key: IVQOVYWBHRSGJI-UHFFFAOYSA-N
SMILES: CCCCCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results