accessibility menu, dialog, popup

UserName

FSOOF

CAS RN 383-29-9

IUPAC Name: 1-fluoro-4-(4-fluorobenzenesulfonyl)benzene
Synonyms: 1-fluoro-4-(4-fluorobenzenesulfonyl)benzene
Molecular Weight (g/mol): 254.25
Molecular Formula: C12H8F2O2S
InChi Key: PLVUIVUKKJTSDM-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)