missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl) Sulfone 99.0+%, TCI America™

Catalog No. D053725G
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053725G 25 g
D053710G 10 g
D0537250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D053725G Supplier TCI America Supplier No. D053725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 383-29-9
Molecular Formula C12H8F2O2S
Molecular Weight (g/mol) 254.25
MDL Number MFCD00000350
InChI Key PLVUIVUKKJTSDM-UHFFFAOYSA-N
Synonym 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
PubChem CID 67842
IUPAC Name 1-fluoro-4-(4-fluorobenzenesulfonyl)benzene
SMILES FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 25 g
Formula Weight 254.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-fluorophenyl) Sulfone