accessibility menu, dialog, popup

UserName

ClOI

CAS RN 38020-81-4

IUPAC Name: 2-iodoacetyl chloride
Synonyms: 2-iodoacetyl chloride
Molecular Weight (g/mol): 204.39
Molecular Formula: C2H2ClIO
InChi Key: BSVMPWANOMFSPR-UHFFFAOYSA-N
SMILES: ClC(=O)CI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)