accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHH3CH3CCH3

CAS RN 37942-07-7

IUPAC Name: 3,5-di-tert-butyl-2-hydroxybenzaldehyde
Synonyms: 3,5-di-tert-butyl-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: RRIQVLZDOZPJTH-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C=O)=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)