accessibility menu, dialog, popup

UserName

CAS RN 37746-78-4

IUPAC Name: ethyl (2E)-4-bromobut-2-enoate
Synonyms: ethyl (2E)-4-bromobut-2-enoate
Molecular Weight (g/mol): 193.04
Molecular Formula: C6H9BrO2
InChi Key: FHGRPBSDPBRTLS-ONEGZZNKSA-N
SMILES: CCOC(=O)\C=C\CBr
Boiling Point 
  • (3)
  • (3)
Grade 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)