accessibility menu, dialog, popup

UserName

CAS RN 375-22-4

IUPAC Name: heptafluorobutanoic acid
Synonyms: heptafluorobutyric acid sodium salt perfluorobutyric acid silver (1+) salt perfluorobutyric acid kyselina heptafluormaselna potassium salt perfluorobutyric acid perfluorobutyric acid, silver (1+) salt
Molecular Weight (g/mol): 214.04
Molecular Formula: C4HF7O2
InChi Key: YPJUNDFVDDCYIH-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (4)
  • (2)
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (7)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (6)
  • (7)