accessibility menu, dialog, popup

UserName

CAS RN 374790-93-9

IUPAC Name: 2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: GWEWQKZABZXLJH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results