CAS RN 374633-31-5
IUPAC Name:
2-bromo-6-methyl-3-nitropyridine
Synonyms:
2-bromo-6-methyl-3-nitropyridine
Molecular Weight (g/mol):
217.02
Molecular Formula:
C6H5BrN2O2
InChi Key:
IHHLJCDGOSZFTB-UHFFFAOYSA-N
SMILES:
CC1=NC(Br)=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2-Bromo-6-methyl-3-nitropyridine, 98%
2-Bromo-6-methyl-3-nitropyridine, 98%
CAS: 374633-31-5 Molecular Formula: C6H5BrN2O2 Molecular Weight (g/mol): 217.022 MDL Number: MFCD03095212 InChI Key: IHHLJCDGOSZFTB-UHFFFAOYSA-N Synonym: 2-bromo-3-nitro-6-methylpyridine,2-bromo-3-nitro-6-picoline,2-bromo-6-methyl-3-nitro-pyridine,2-bromo-3-nitro-6-methyl pyridine,pubchem5954,acmc-209it3,ksc496c9p,6-bromo-5-nitro-2-picoline,pyridine, 2-bromo-6-methyl-3-nitro,2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate PubChem CID: 24728013 IUPAC Name: 2-bromo-6-methyl-3-nitropyridine SMILES: CC1=NC(=C(C=C1)[N+](=O)[O-])Br