CAS RN 3736-81-0
CAS RN 3736-81-0
IUPAC Name:
4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate
Synonyms:
diloxanide|diloxanide furoate|entamide furoate|miforon|amebiazol|furentomin|histomibal|diclofurazol|dichlofurazol|diloxanid furoate
Molecular Weight (g/mol):
328.15
Molecular Formula:
C14H11Cl2NO4
InChi Key:
BDYYDXJSHYEDGB-UHFFFAOYSA-N
SMILES:
CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1
1
–
1
of
1
results
1
Thermo Scientific™ Diloxanide (furoate), 97%
CAS: 3736-81-0 Molecular Formula: C14H11Cl2NO4 Molecular Weight (g/mol): 328.15 MDL Number: MFCD00083302 InChI Key: BDYYDXJSHYEDGB-UHFFFAOYSA-N IUPAC Name: 4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate SMILES: CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1