accessibility menu, dialog, popup

UserName

CAS RN 373-57-9

IUPAC Name: trifluoroborane
Synonyms: boron trifluoride|boron fluoride|bortrifluorid
Molecular Weight (g/mol): 67.81
Molecular Formula: BF3
InChi Key: WTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILES: FB(F)F
Molecular Weight (g/mol) 
  • (10)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (5)
  • (10)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (4)