missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Boiling Point | 65.0°C |
| Chemical Name or Material | Boron trifluoride |
| Melting Point | -98.0°C |
| CAS | 373-57-9, 67-56-1 |
| Quantity | 800 mL |
| Flash Point | 4°C |
| Packaging | AcroSeal™ Glass bottle |
| Specific Gravity | 0.87 |
| Density | 0.8700g/mL |
| Molecular Formula | BF3 |
| Show More |
RUO – Research Use Only