missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride - Methanol Reagent (10-20%) [for Esterification] (1mL*10), TCI America™

Catalog No. X0036SET
Change view
Click to view available options
Quantity:
Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0036SET Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X0036SET Supplier TCI America Supplier No. X00361SET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 373-57-9
Molecular Formula BF3
Molecular Weight (g/mol) 67.81
MDL Number MFCD00011316
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
PubChem CID 11062313
IUPAC Name trifluoroborane
SMILES FB(F)F

Specifications

Color Yellow
Physical Form Liquid
Quantity Set
UN Number 2924
Chemical Name or Material Boron Trifluoride - Methanol Reagent (10-20%) [for Esterification] (1mL*10)
EINECSNumber (2)-2112
RTECSNumber ED2275000
TSCA Yes