missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride, 14% in Methanol, Spectrum™ Chemical

Catalog No. 18601063
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601063 1 L Amber Glass Bottle
18601060 100 mL Amber Glass Bottle
18601061 250 mL Amber Glass Bottle
18601062 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601063 Supplier Spectrum Chemical Mfg Cor Supplier No. B26441LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B2644, 373-57-9, CH4BF3O

Spectrum™ Chemical Boron Trifluoride, 14 Percent in Methanol is used as a reagent in organic chemistry most often as a Lewis acid. It is known to be a versatile building block in several other boron compounds. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 373-57-9
Molecular Formula BF3
Molecular Weight (g/mol) 67.81
MDL Number MFCD00011316
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
IUPAC Name trifluoroborane
SMILES FB(F)F

Specifications

CAS Max % 100%
Physical Form Liquid
Quantity 1 L
UN Number UN3286
Grade Ungraded
Concentration or Composition (by Analyte or Components) 14%
Packaging Amber Glass Bottle
Chemical Name or Material Boron Trifluoride