accessibility menu, dialog, popup

UserName

NH2HNH2NOOHO

CAS RN 372-75-8

IUPAC Name: 2-amino-5-(carbamoylamino)pentanoic acid
Synonyms: citrullin N(5)-carbamoyl-DL-ornithine citrulina dl-citrulline N(5)-(aminocarbonyl)ornithine
Molecular Weight (g/mol): 175.19
Molecular Formula: C6H13N3O3
InChi Key: RHGKLRLOHDJJDR-UHFFFAOYNA-N
SMILES: NC(CCCNC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (9)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (3)
  • (2)
  • (4)
  • (4)
  • (9)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)