CAS RN 37107-81-6
IUPAC Name:
5-ethenyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
5-ethenyl-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol):
138.13
Molecular Formula:
C6H6N2O2
InChi Key:
ZRYZBEQILKESAW-UHFFFAOYSA-N
SMILES:
C=CC1=CNC(=O)NC1=O
1
–
1
of
1
results
1
5-Vinyluracil, 97%
CAS: 37107-81-6 Molecular Formula: C6H6N2O2 Molecular Weight (g/mol): 138.13 MDL Number: MFCD00913267 InChI Key: ZRYZBEQILKESAW-UHFFFAOYSA-N Synonym: 5-vinyluracil,5-ethenyl-2,4 1h,3h-pyrimidinedione,5-ethenyluracil,5-vinyl-1h-pyrimidine-2,4-dione,5-vinylpyrimidine-2,4 1h,3h-dione,5-ethenyl-1,3-dihydropyrimidine-2,4-dione,5-ethenylpyrimidine-2,4-diol,acmc-1aif7,2,4 1h,3h-pyrimidinedione,5-ethenyl PubChem CID: 99305 IUPAC Name: 5-ethenyl-1H-pyrimidine-2,4-dione SMILES: C=CC1=CNC(=O)NC1=O