accessibility menu, dialog, popup

UserName

CH3OHNSHN

CAS RN 37052-78-1

IUPAC Name: 5-methoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-methoxy-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 180.23
Molecular Formula: C8H8N2OS
InChi Key: KOFBRZWVWJCLGM-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=S)NC2=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)