accessibility menu, dialog, popup

UserName

OHNNONHN

CAS RN 370-81-0

IUPAC Name: N',N'-dicyclohexylideneethanedihydrazide
Synonyms: N',N'-dicyclohexylideneethanedihydrazide
Molecular Weight (g/mol): 278.36
Molecular Formula: C14H22N4O2
InChi Key: DSRJIHMZAQEUJV-UHFFFAOYSA-N
SMILES: O=C(NN=C1CCCCC1)C(=O)NN=C1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)