missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid bis(cyclohexylidenehydrazide), 98%

Catalog No. AC154421000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154421000 100 g
AC154420250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154421000 Supplier Thermo Scientific Chemicals Supplier No. 154421000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 370-81-0 | C14H22N4O2 | 278.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 370-81-0
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.34
MDL Number MFCD00001659
InChI Key DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonym cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
IUPAC Name N,N'-bis(cyclohexylideneamino)oxamide
SMILES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Melting Point 208.0°C to 214.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 07, IV, 25
Formula Weight 278.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Oxalic acid bis(cyclohexylidenehydrazide), 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-729-2
RTECSNumber RO2520000
TSCA TSCA

RUO – Research Use Only