accessibility menu, dialog, popup

UserName

CAS RN 3694-57-3

IUPAC Name: 1-(isothiocyanatomethyl)-4-methoxybenzene
Synonyms: 1-(isothiocyanatomethyl)-4-methoxybenzene
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: IMFQYAJJXFXVMM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN=C=S)C=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)