CAS RN 3680-02-2
IUPAC Name:
methanesulfonylethene
Synonyms:
methyl vinyl sulfone
Molecular Weight (g/mol):
106.14
Molecular Formula:
C3H6O2S
InChi Key:
WUIJTQZXUURFQU-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C=C
1
–
1
of
1
results
1
Promotions Available
Methyl vinyl sulfone, 95%, stabilized
Methyl vinyl sulfone, 95%, stabilized
CAS: 3680-02-2 Molecular Formula: C3H6O2S Molecular Weight (g/mol): 106.14 MDL Number: MFCD00007567 InChI Key: WUIJTQZXUURFQU-UHFFFAOYSA-N SMILES: CS(=O)(=O)C=C