accessibility menu, dialog, popup

UserName

CAS RN 36637-44-2

IUPAC Name: 2-[4-(bromomagnesio)phenoxy]oxane
Synonyms: 2-[4-(bromomagnesio)phenoxy]oxane
Molecular Weight (g/mol): 281.43
Molecular Formula: C11H13BrMgO2
InChi Key: OHPUQUBYKZJOLY-UHFFFAOYNA-M
SMILES: Br[Mg]C1=CC=C(OC2CCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results