accessibility menu, dialog, popup

UserName

CH3SOONC

CAS RN 36635-66-2

IUPAC Name: 1-isocyano(phenyl)methanesulfonyl-4-methylbenzene
Synonyms: 1-isocyano(phenyl)methanesulfonyl-4-methylbenzene
Molecular Weight (g/mol): 271.33
Molecular Formula: C15H13NO2S
InChi Key: KIJCBVOPJSHLBI-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results