accessibility menu, dialog, popup

UserName

CAS RN 36635-61-7

IUPAC Name: 1-isocyanomethanesulfonyl-4-methylbenzene
Synonyms: TosMIC
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: CFOAUYCPAUGDFF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)