accessibility menu, dialog, popup

UserName

CAS RN 3647-69-6

IUPAC Name: hydrogen 4-(2-chloroethyl)morpholine chloride
Synonyms: hydrogen morpholine, 4-(2-chloroethyl)- chloride
Molecular Weight (g/mol): 186.08
Molecular Formula: C6H13Cl2NO
InChi Key: NBJHDLKSWUDGJG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCCN1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)