accessibility menu, dialog, popup

UserName

CAS RN 3634-83-1

IUPAC Name: 1,3-bis(isocyanatomethyl)benzene
Synonyms: 1,3-bis(isocyanatomethyl)benzene
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: RTTZISZSHSCFRH-UHFFFAOYSA-N
SMILES: O=C=NCC1=CC(CN=C=O)=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results