accessibility menu, dialog, popup

UserName

ClMgBr

CAS RN 36229-42-2

IUPAC Name: bromo(3-chlorophenyl)magnesium
Synonyms: bromo(3-chlorophenyl)magnesium
Molecular Weight (g/mol): 215.76
Molecular Formula: C6H4BrClMg
InChi Key: ZIYRUWPTUPFRQU-UHFFFAOYSA-M
SMILES: ClC1=CC([Mg]Br)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)