accessibility menu, dialog, popup

UserName

CAS RN 35941-71-0

HClHONNONClSO

CAS RN 35941-71-0

IUPAC Name: hydrogen 5-chloro-3-{2-[4-(2-hydroxyethyl)piperazin-1-yl]-2-oxoethyl}-2,3,3a,7a-tetrahydro-1,3-benzothiazol-2-one chloride
Synonyms: hydrogen 5-chloro-3-{2-[4-(2-hydroxyethyl)piperazin-1-yl]-2-oxoethyl}-3a,7a-dihydro-1,3-benzothiazol-2-one chloride
Molecular Weight (g/mol): 394.31
Molecular Formula: C15H21Cl2N3O3S
InChi Key: XJTXBQFPQSKSPV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OCCN1CCN(CC1)C(=O)CN1C2C=C(Cl)C=CC2SC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results