accessibility menu, dialog, popup

UserName

CAS RN 358727-55-6

IUPAC Name: 7-(2,4-dihydroxy-3-{4,4,10,10-tetramethyl-6-oxido-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-7-yl}cyclobuta-1,3-dien-1-yl)-4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-6-olate
Synonyms: 7-(2,4-dihydroxy-3-{4,4,10,10-tetramethyl-6-oxido-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-7-yl}cyclobuta-1,3-dien-1-yl)-4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-6-olate
Molecular Weight (g/mol): 568.76
Molecular Formula: C36H44N2O4
InChi Key: ZVJJZBXHRPLGNI-UHFFFAOYSA-L
SMILES: CC1(C)CCN2CCC(C)(C)C3=C([O-])C(=CC1=C23)C1=C(O)C(=C1O)C1=CC2=C3N(CCC2(C)C)CCC(C)(C)C3=C1[O-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results