accessibility menu, dialog, popup

UserName

H3CCH3(S)NHOOOHO

CAS RN 35661-60-0

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
Synonyms: fmoc-L-leucine
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: CBPJQFCAFFNICX-TWYLJJHKNA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)