accessibility menu, dialog, popup

UserName

CAS RN 3561-67-9

IUPAC Name: {[(phenylsulfanyl)methyl]sulfanyl}benzene
Synonyms: {[(phenylsulfanyl)methyl]sulfanyl}benzene
Molecular Weight (g/mol): 232.36
Molecular Formula: C13H12S2
InChi Key: ZHUPZVIALZHGGP-UHFFFAOYSA-N
SMILES: C(SC1=CC=CC=C1)SC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)