missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylthio)methane 97.0+%, TCI America™

Catalog No. B14445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14445G 5 g
B144425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14445G Supplier TCI America Supplier No. B14445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3561-67-9
Molecular Formula C13H12S2
Molecular Weight (g/mol) 232.359
MDL Number MFCD00003067
InChI Key ZHUPZVIALZHGGP-UHFFFAOYSA-N
Synonym bis phenylthio methane, methylenebis thio bisbenzene, bis phenyithio methane, formaldehyde diphenyl mercaptal, 1,1'-methylenebis thio bisbenzene, phenylsulfanyl methyl sulfanyl benzene, 1-phenylthio methyl thio benzene, bis thiophenoxy methane
PubChem CID 77097
IUPAC Name phenylsulfanylmethylsulfanylbenzene
SMILES C1=CC=C(C=C1)SCSC2=CC=CC=C2

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 232.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(phenylthio)methane