CAS RN 355423-58-4
IUPAC Name:
6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms:
6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol):
246.04
Molecular Formula:
C8H5BrFNO2
InChi Key:
AVHJMSMGASUDIT-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=C2NC(=O)COC2=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 96%, Thermo Scientific Chemicals
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one, 96%, Thermo Scientific Chemicals
CAS: 355423-58-4 Molecular Formula: C8H5BrFNO2 Molecular Weight (g/mol): 246.04 MDL Number: MFCD20441932 InChI Key: AVHJMSMGASUDIT-UHFFFAOYSA-N Synonym: 6-bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one,6-bromo-7-fluoro-2h-benzo b 1,4 oxazin-3 4h-one,2h-1,4-benzoxazin-3 4h-one, 6-bromo-7-fluoro,acmc-209ihm,6-bromo-7-fluoro-4h-benzo 1,4 oxazin-3-one,6-bromo-7-fluoro-2h-1,4-benzoxazin-3 4h-one,6-bromo-7-fluoro-3,4-dihydro-2h-1,4-benzoxazin-3-one PubChem CID: 23574320 IUPAC Name: 6-bromo-7-fluoro-4H-1,4-benzoxazin-3-one SMILES: FC1=C(Br)C=C2NC(=O)COC2=C1