accessibility menu, dialog, popup

UserName

H3COOOCH3Br

CAS RN 35450-37-4

IUPAC Name: methyl 3-bromo-4-methoxybenzoate
Synonyms: methyl 3-bromo-4-methoxybenzoate
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: ZREVPFANJBZHEU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(OC)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)