accessibility menu, dialog, popup

UserName

CAS RN 354-37-0

IUPAC Name: trifluoroethanimidamide
Synonyms: trifluoroethanimidamide
Molecular Weight (g/mol): 112.06
Molecular Formula: C2H3F3N2
InChi Key: NITMACBPVVUGOJ-UHFFFAOYSA-N
SMILES: NC(=N)C(F)(F)F
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)