CAS RN 35051-49-1
IUPAC Name:
ethyl 2-(diethoxyphosphoryl)pentanoate
Synonyms:
ethyl 2-(diethoxyphosphoryl)pentanoate
Molecular Weight (g/mol):
266.27
Molecular Formula:
C11H23O5P
InChi Key:
BUPVIVDUPRDDFI-UHFFFAOYNA-N
SMILES:
CCCC(C(=O)OCC)P(=O)(OCC)OCC
1
–
1
of
1
results
1
Triethyl 2-phosphonopentanoate, 98%
CAS: 35051-49-1 Molecular Formula: C11H23O5P Molecular Weight (g/mol): 266.274 MDL Number: MFCD00015163 InChI Key: BUPVIVDUPRDDFI-UHFFFAOYSA-N Synonym: triethyl 2-phosphonopentanoate,ethyl 2-diethoxyphosphoryl pentanoate,pentanoic acid, 2-diethoxyphosphinyl-, ethyl ester,valeric acid, 2-diethylphosphono-, ethyl ester,alpha-carbethoxybutylphosphonic acid, diethyl ester,triethyl 2-phosphonovalerate,phosphonic acid, alpha-carbethoxybutyl-, diethyl ester,ethyl 2-diethoxyphosphoryl pentanoate #,diethyl 1-ethoxycarbonyl butylphosphonate PubChem CID: 98244 IUPAC Name: ethyl 2-diethoxyphosphorylpentanoate SMILES: CCCC(C(=O)OCC)P(=O)(OCC)OCC